1,3-Dipalmitolein ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

32165325MG
100290-318EA 448.54 USD
100290-318
1,3-Dipalmitolein ≥99%
1,3-Dipalmitolein

Store in a cool, dry place

MDL Number: MFCD00673498
CAS Number: 113728-10-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e9e1dd788f6f73e86799971f92585ef726c954a&vid=3e9e1dd788f6f73e86799971f92585ef726c954a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR