Monononadecanoin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2,3-dihydroxypropyl nonadecanoate

31190050MG
100290-220EA 223.62 USD
100290-220
Monononadecanoin ≥99%
Monononadecanoin

Store in a cool, dry place

Formula: C₂₂H₄₄O₄
MDL Number: MFCD00673486
CAS Number: 62927-07-5
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR