VWR® Isolator Tool Cover
Supplier: Micronova
ITCAA-9
10029-772CS 619.09 USD
10029-772
VWR® Isolator Tool Cover
Mops
An elasticized microfiber slip cover for use with the QDIT/QPIT series mop adapters.
  • Microfiber
  • Lot numbers printed on each bag and each case
Designed for isolators, bio-hazard and containment hoods.
Certifications: ISO 9001:2008 Facility.
Packaging: Each mop head single bagged and packaged 50 mops per poly lined case.