General Purpose Heating Circulators, Grant Instruments
Supplier: GRANT USA INC.
T100 US TC120 US T100-ST5 US T100-ST12 US T100-ST18 US T100-ST26 US T100-ST38 US TC120-ST5 US TC120-ST12 US TC120-ST18 US TC120-ST26 US TC120-ST38 US
89260-838EA 1543 USD
89260-838 89260-840 10029-438 10029-440 10029-442 10029-444 10029-446 10029-448 10029-450 10029-452 10029-454 10029-456 10029-458 10029-460 10029-462 10029-464 10029-466 10029-468 10029-502 10029-504
General Purpose Heating Circulators, Grant Instruments
Circulating Thermostats Closed Circulator Systems Heating Circulators
The Optima™ range of general purpose thermostats can be combined with plastic baths (5L, 12L, 18L), premium stainless steel baths (5L, 12L, 18L, 26L, 38L) or refrigeration units (3 units available)
Units feature a bright, easy to view four digit display, three programmable temperature presets, low liquid protection, fixed over-temperature cut-out, visual alarm, countdown timer (TC120) and user calibration facility for optimum accuracy.
T100 temperature range: Ambient +5 to 100 °C. Settable range 0 to 100 °C (operation below ambient requires accessory cooling). TC120 temperature range: Ambient +5 to 120 °C. Settable range −20 to +120 °C (operation below ambient requires accessory cooling). TC120 temperature range: Ambient +5 to 120 °C. Settable range −20 to +120 °C (operation below ambient requires accessory cooling). Stability: ±0,05 °C at 70 °C. TC120 has an audible and visual alarm, an adjustable over-temperature cut-out and 16L/min external circulation. Drain tap with all tanks greater than 12L.
Digital microprocessor controlled T100 and TC120 thermostats feature an easy to use rotary dial and two function keys for quick temperature setting and menu navigation.
Certifications: CE Certified.
Ordering information: Many accessories available including; lids, racks, shelves and refrigeration units.