Shipper, EAS Frozen, Therapak®
Supplier: Therapak, LLC
56885
10029-420CS 127 USD
10029-420
Shipper, EAS Frozen, Therapak®
Mailers and Shippers
EAS shipping systems for frozen specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and transport box (OD: 11 x 9 x 11”) with a Class 9 Dry Ice label, making it ideal for overnight shipments.
Includes specimen bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label, and cushioning material. Add sample tubes and dry ice to ship.