Shipper, Multisample EAS Ambient, Therapak®
Supplier: Therapak, LLC
56883
10029-416CS 107.67 USD
10029-416
Shipper, Multisample EAS Ambient, Therapak®
Mailers and Shippers
Large capacity ambient shipping system accommodates multiple specimen transport bags for consolidation at collection site and shipping to testing laboratory
Features a pop-up style box that is easy to assemble. Easy-fold flaps on both ends of the box enable the shipper to easily slide the included specimen bag and absorbent containing sample tubes inside the box prior to securing the flaps for shipment. Includes an Exempt Animal Specimen label and biohazard symbol label.
System includes a large transport box (ID: 11 x 10.5 x 4"; OD: 11.25 x 10.75 x 4.25") with easy lock flaps on both ends of the box, a 13 x 18" biohazard transport bag, and 12 x 12" absorbent sheet. Holds up to ten 6 x 9" specimen transport bags.