Shipper, EAS Ambient for 4 Tubes, Therapak®
Supplier: Therapak, LLC
56881
10029-412CS 258.84 USD
10029-412
Shipper, EAS Ambient for 4 Tubes, Therapak®
Mailers and Shippers
Ambient shipping system features an Aqui-Pak™ segmented absorbent pouch to hold up to four standard blood or urine tubes.
Shipped with pre-assembled boxes, these transport shippers also include: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label and cushioning material where applicable.