Shipper, EHS Frozen, Therapak®
Supplier: Therapak, LLC
56877
10029-404CS 89.47 USD
10029-404
Shipper, EHS Frozen, Therapak®
Mailers and Shippers
EHS shipping systems for frozen specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and box (OD: 11 x 9 x 11”) with a Class 9 Dry Ice label, making it ideal for overnight shipments.
Includes specimen bag, absorbent material, Exempt Human Specimen label, biohazard symbol label, and cushioning material. Add sample tubes and dry ice to ship.