Shipper, EHS Ambient for 4 Tubes, Therapak®
Supplier: Therapak, LLC
56873
10029-396CS 278.01 USD
10029-396
Shipper, EHS Ambient for 4 Tubes, Therapak®
Mailers and Shippers
Ambient shipping system features: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, Aqui-Pak™ segmented absorbent pouch to hold up to four standard blood or urine tubes, Exempt Human Specimen label, biohazard symbol label and cushioning material where applicable.