Shipper, EHS Ambient, Therapak®
Supplier: Therapak, LLC
56872
10029-394CS 219.21 USD
10029-394
Shipper, EHS Ambient, Therapak®
Mailers and Shippers
Ambient shipping system is designed for shipping one standard tube or larger specimen containers (30 mL to 60 mL) of liquid specimen
System features: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Human Specimen label, biohazard symbol label and cushioning material where applicable.