Shipper, Category B Refrigerated, 2...8 °C, Therapak®
Supplier: Therapak, LLC
38028
10029-326CS 413.97 USD
10029-326
Shipper, Category B Refrigerated, 2...8 °C, Therapak®
Mailers and Shippers
Refrigerated Shipping System has been field tested for 2 - 8° C temperature maintenance for 48 hours
System includes: 12 x 12 x 12” 200# corrugated outer box, EPS cooler with 21/2” all around wall thickness, ambient gel wrap, 95kPa specimen transport bag with absorbent to hold up to six specimen tubes, refrigerant packs, list of contents card, filler material, and required labels for IATA PI650 marking and compliance. Packing Instructions are included to facilitate use for safe and compliant shipping.