Frozen Diagnostic Shipping System, Therapak®
Supplier: Therapak, LLC
37807
10029-270CS 308.64 USD
10029-270
Frozen Diagnostic Shipping System, Therapak®
Mailers and Shippers
Frozen Shipping System includes Aqui-Pak 6 bay absorbent pouch, 12.25x15.25" 95 kPa Biohazard Bag, Box for four medium coolers, Bubble Wrap, Dry Ice Label, Biohazard Label, Frozen Specimen Label, Double Up Arrow, UN3373 Biological Substance, Cat B Label, List of Contents Card, Medium Foam Cooler, Medium Cooler Box.