Shipper, Category B Ambient, 95 kPa Bag, Tube Shuttle, Therapak®
Supplier: Therapak, LLC
37802
10029-266CS 282.53 USD
10029-266
Shipper, Category B Ambient, 95 kPa Bag, Tube Shuttle, Therapak®
Mailers and Shippers
The ambient transport system for blood and urine tube transport includes 95kPa bag, TearTech tube shuttle, absorbent sheet, cushioning material, list of contents card, biohazard symbol label, Biological Substance Category B label, and medium transport box.
Holds up to six tubes.