Shipper, Category B Ambient, 95 kPa Medium Canister, Therapak®
Supplier: Therapak, LLC
37804
10029-258CS 282.08 USD
10029-258
Shipper, Category B Ambient, 95 kPa Medium Canister, Therapak®
Mailers and Shippers
The ambient transport system for blood and urine tube transport includes medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, cushioning material, list of contents card, biohazard symbol label, Biological Substance Category B label, and medium transport box.
Holds up to six tubes.