1,1,1,2,2-Pentafluoro-3-oxopentane ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Pentafluoroethyl ethyl ketone

272131GM
100289-760EA 184.38 USD
100289-760
1,1,1,2,2-Pentafluoro-3-oxopentane ≥97%
1,1,1,2,2-Pentafluoro-3-oxopentane
Formula: C₅H₅F₅O
MW: 176.09 g/mol
MDL Number: MFCD00053172
CAS Number: 378-72-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abfeb3665084e7fb5fe499c3cf87242ad5ab1796&vid=abfeb3665084e7fb5fe499c3cf87242ad5ab1796&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR