(±)-3-Fluoro-α-methylbenzylamine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(3-Fluorophenyl)ethanamine

271501GM
100289-574EA 289 USD
100289-574
(±)-3-Fluoro-α-methylbenzylamine ≥97%
(±)-3-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD04038308
CAS Number: 74788-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24980f6494c141165db871b1419fb60c213f04ee&vid=24980f6494c141165db871b1419fb60c213f04ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR