2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

266731GM
100288-952EA 107.23 USD
100288-952
2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%
2-Bromo-4'-(difluoromethoxy)acetophenone
Formula: C₉H₇BrF₂O₂
Melting Pt: 63…68 °C
MDL Number: MFCD00052200
CAS Number: 141134-24-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=786497e3ab1bf8c55c35d9330431c3a44ef4499c&vid=786497e3ab1bf8c55c35d9330431c3a44ef4499c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR