3,5-Difluoro-o-phenylenediamine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

265481GM
100288-858EA 104.62 USD
100288-858
3,5-Difluoro-o-phenylenediamine ≥98%
3,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
Melting Pt: 48…50 °C
MDL Number: MFCD00973899
CAS Number: 2369-29-1
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ccdfca404620ff3abd192690bb66e9d8f13ffe9&vid=1ccdfca404620ff3abd192690bb66e9d8f13ffe9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR