4-Chloro-6-(trifluoromethyl)quinoline ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

265081GM
100288-832EA 176.54 USD
100288-832
4-Chloro-6-(trifluoromethyl)quinoline ≥98%
4-Chloro-6-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00153106
CAS Number: 49713-56-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac6565af57746b2062e3a0d846b96eae75bbc292&vid=ac6565af57746b2062e3a0d846b96eae75bbc292&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR