3-(3-Trifluoromethylphenyl)propionic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid

2646055GM
100288-688EA 115.08 USD
100288-688
3-(3-Trifluoromethylphenyl)propionic acid ≥98%
3-(3-Trifluoromethylphenyl)propionic acid
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Melting Pt: 29…31 °C
MDL Number: MFCD00090487
CAS Number: 585-50-2
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9383bb63139e5239ff3fd1d5321e11cdf647bec0&vid=9383bb63139e5239ff3fd1d5321e11cdf647bec0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR