3-(Trifluoromethoxy)-DL-phenylglycine

Supplier: INDOFINE CHEMICAL COMPANY MS

2645371GM
100288-560EA 503.46 USD
100288-560
3-(Trifluoromethoxy)-DL-phenylglycine
3-(Trifluoromethoxy)-DL-phenylglycine
Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
MDL Number: MFCD01631460
CAS Number: 261952-23-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0937b1de543c0d8e5ed55a23cedb1eb281af0a18&vid=0937b1de543c0d8e5ed55a23cedb1eb281af0a18&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955d92f51ca8c1f74d76e4aa8bc820dad44e7d6f&vid=955d92f51ca8c1f74d76e4aa8bc820dad44e7d6f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR