1,1,1-Trifluoroiodoethane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

26407025GM
100288-012EA 145.15 USD
100288-012
1,1,1-Trifluoroiodoethane ≥99%
1,1,1-Trifluoroiodoethane
Formula: C₂H₂F₃I
Boiling Pt: 53…55 °C
MDL Number: MFCD00001082
CAS Number: 353-83-3
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa16d4a096ebadc253e9ce5a1db07dafc932afb0&vid=fa16d4a096ebadc253e9ce5a1db07dafc932afb0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR