3-Fluoro-4-nitrobenzoic acid ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2637905GM
100287-842EA 104.62 USD
100287-842
3-Fluoro-4-nitrobenzoic acid ≥97%
3-Fluoro-4-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11
Melting Pt: 174…175 °C
MDL Number: MFCD01862092
CAS Number: 403-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1babd1271eee80d34d3ba88954aa63d1e6f86046&vid=1babd1271eee80d34d3ba88954aa63d1e6f86046&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR