1-(Difluoromethoxy)-3-nitrobenzene ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-(Difluoromethoxy)nitrobenzene

2632785GM
100287-392EA 104.62 USD
100287-392
1-(Difluoromethoxy)-3-nitrobenzene ≥98%
1-(Difluoromethoxy)-3-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 240…242 °C
MDL Number: MFCD03407974
CAS Number: 22236-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d37f66d6fc14d9a7262af11539c2bb500b2c95&vid=03d37f66d6fc14d9a7262af11539c2bb500b2c95&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR