2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2631041GM
100287-306EA 160.85 USD
100287-306
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone ≥97%
2,3-Diamino-6-(trifluoromethyl)-4-(3H)-pyrimidinone
Formula: C₅H₅F₃N₄O
MW: 194.12 g/mol
Melting Pt: 226…228 °C
MDL Number: MFCD00068010
CAS Number: 95095-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e6c8b179d92f53fb760a891a4c5b8da21cea04&vid=91e6c8b179d92f53fb760a891a4c5b8da21cea04&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR