4-(Trifluoromethoxy)-1,2-phenylenediamine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2631031GM
100287-302EA 185.69 USD
100287-302
4-(Trifluoromethoxy)-1,2-phenylenediamine ≥97%
4-(Trifluoromethoxy)-1,2-phenylenediamine

  • Pale yellow oil
  • Store in a cool, dry place

Formula: C₇H₇F₃N₂O
MDL Number: MFCD03094278
CAS Number: 658-89-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d1bd3174338e40ef68894973f5e947eb18a6e0d&vid=4d1bd3174338e40ef68894973f5e947eb18a6e0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR