3-Chloro-2-fluorophenylhydrazine hydrochloride

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (3-Chloro-2-fluorophenyl)hydrazine hydrochloride

2628685GM
100286-992EA 150.38 USD
100286-992
3-Chloro-2-fluorophenylhydrazine hydrochloride
3-Chloro-2-fluorophenylhydrazine hydrochloride
Formula: C₆H₇Cl₂FN₂
MW: 197.0400 g/mol
MDL Number: MFCD03094280
CAS Number: 517920-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab004c6985928450bc0e0beffeecea393601c519&vid=ab004c6985928450bc0e0beffeecea393601c519&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR