1-Chloro-1,2-difluoroethylene ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2627555GM
100286-888EA 804.23 USD
100286-888
1-Chloro-1,2-difluoroethylene ≥97%
1-Chloro-1,2-difluoroethylene
Formula: C₂HClF₂
MDL Number: MFCD01726891
CAS Number: 359-04-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a118a1f1cea1fdcb7eef2ff92bd21610b2d31ec&vid=2a118a1f1cea1fdcb7eef2ff92bd21610b2d31ec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR