1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

262355GM
100286-618EA 411.92 USD
100286-618
1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene ≥97%
1-Bromo-2-chloro-1,1,2-trifluoro-3-pentene
Formula: C₅H₅BrClF₃
MW: 237.45 g/mol
MDL Number: MFCD00153682
CAS Number: 356-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca03336623c597070bd1be7577849cc1991850f9&vid=ca03336623c597070bd1be7577849cc1991850f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR