4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

2622221GM
100286-506EA 346.54 USD
100286-506
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether
Formula: C₁₄H₉F₃N₂S
Melting Pt: 78…80 °C
MDL Number: MFCD00793554
CAS Number: 320421-51-0
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a5a4814786727689f824f37f6f93980251b9b6b&vid=1a5a4814786727689f824f37f6f93980251b9b6b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR