ω-Bromo-3-fluoroacetophenone ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2610065GM
100285-678EA 188.31 USD
100285-678
ω-Bromo-3-fluoroacetophenone ≥97%
ω-Bromo-3-fluoroacetophenone
Formula: C₈H₆BrFO
Melting Pt: 32…37 °C
MDL Number: MFCD00109756
CAS Number: 53631-18-8
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f1db76be8886303ae993a0c5518ad7213ade3fa&vid=4f1db76be8886303ae993a0c5518ad7213ade3fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR