Methyl-3-hydroxyhexanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Methyl 3-hydroxyhexanoate

24060325MG
100285-392EA 451.15 USD
100285-392
Methyl-3-hydroxyhexanoate ≥98%
Methyl 3-hydroxyhexanoate
Formula: C₇H₁₄O₃
MW: 146.1843 g/mol
MDL Number: MFCD00083583
CAS Number: 21188-58-9
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ba18f423a2761a8eeb1de40a6a6253a5af87e8a&vid=8ba18f423a2761a8eeb1de40a6a6253a5af87e8a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR