2-(Trifluoromethyl)benzophenone ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Phenyl(2-(trifluoromethyl)phenyl)methanone

215615GM
100285-356EA 111.15 USD
100285-356
2-(Trifluoromethyl)benzophenone ≥98%
2-(Trifluoromethyl)benzophenone
Formula: C₁₄H₉F₃O
MW: 250.22
Melting Pt: 62
MDL Number: MFCD00000377
CAS Number: 727-99-1
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=436bccd8fc25a6cd0e2205c145bc08d81072099b&vid=436bccd8fc25a6cd0e2205c145bc08d81072099b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aff872a45c92c6b364de36ea77a7736221fb939a&vid=aff872a45c92c6b364de36ea77a7736221fb939a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR