4-Fluoro-3-hydroxybenzoic acid ≥97%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

213305GM
100285-136EA 156.92 USD
100285-136
4-Fluoro-3-hydroxybenzoic acid ≥97%, crystalline powder
4-Fluoro-3-hydroxybenzoic acid

  • pale orange crystalline powder
  • Store in a cool, dry place

Formula: C₇H₅FO₃
Melting Pt: 214…220 °C
MDL Number: MFCD00269641
CAS Number: 51446-31-2
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d7cc12e636498303f76b3a8235a9bb8ad2136a&vid=55d7cc12e636498303f76b3a8235a9bb8ad2136a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR