3-Fluoro-4-hydroxybenzaldehyde ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

213295GM
100285-134EA 119 USD
100285-134
3-Fluoro-4-hydroxybenzaldehyde ≥98%
3-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
Melting Pt: 121…124 °C
MDL Number: MFCD00016626
CAS Number: 405-05-0
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff730122df572afe8938a7362a5cbb68e12ba1f2&vid=ff730122df572afe8938a7362a5cbb68e12ba1f2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR