Methyl pristanate

Supplier: INDOFINE CHEMICAL COMPANY MS

2115005MG
100285-024EA 627.69 USD
100285-024
Methyl pristanate
Methyl pristanate

Store in a cool, dry place

Formula: C₂₀H₄₀O₂
MDL Number: MFCD04040009
CAS Number: 1001-80-5
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29ac17f8a81db2948992bf344f31d4090ebeda50&vid=29ac17f8a81db2948992bf344f31d4090ebeda50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR