1-Bromo-2-fluorobenzene ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Bromofluorobenzene

21030100GM
100284-838EA 125.54 USD
100284-838
1-Bromo-2-fluorobenzene ≥99%
1-Bromo-2-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155
MDL Number: MFCD00000282
CAS Number: 1072-85-1
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faa2d12a5f3c138b68ebd6974a83b9866a2dd381&vid=faa2d12a5f3c138b68ebd6974a83b9866a2dd381&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR