2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4,4'-Diiodo-2,2'-bis(trifluoromethyl)-1,1'-biphenyl

21023250MG
100284-828EA 351.77 USD
100284-828
2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl ≥97%
2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl
Formula: C₁₄H₆F₆I₂
MW: 541.99 g/mol
MDL Number: MFCD00236673
CAS Number: 89803-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeb11d3746a376536f9a2637e4f186817144a06f&vid=aeb11d3746a376536f9a2637e4f186817144a06f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=026b783159e5921a1de7c87652c9c7020122fe6c&vid=026b783159e5921a1de7c87652c9c7020122fe6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR