3,3'-Bis(trifluoromethyl)benzhydrol ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

210171GM
100284-814EA 156.92 USD
100284-814
3,3'-Bis(trifluoromethyl)benzhydrol ≥97%
3,3'-Bis(trifluoromethyl)benzhydrol
Formula: C₁₅H₁₀F₆O
MDL Number: MFCD00039228
CAS Number: 1598-89-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8696f59f9e701d0a269286cbe23783901fafaf45&vid=8696f59f9e701d0a269286cbe23783901fafaf45&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=842471ba66220396f8b833baa8278ec5de15c204&vid=842471ba66220396f8b833baa8278ec5de15c204&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR