1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2,2'-(Benzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

210145GM
100284-810EA 146.46 USD
100284-810
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥97%
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Formula: C₁₂H₆F₁₂O₂
MW: 410.16
MDL Number: MFCD00042090
CAS Number: 802-93-7
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1d84991d817125d60f9ef84dfc55a08ec5888dd&vid=a1d84991d817125d60f9ef84dfc55a08ec5888dd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR