Methyl heptacosanoate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

20270025MG
100284-788EA 612 USD
100284-788
Methyl heptacosanoate ≥99%
Methyl heptacosanoate

Store in a cool, dry place

Formula: C₂₈H₅₆O₂
MW: 424.754 g/mol
MDL Number: MFCD00057798
CAS Number: 55682-91-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cb223000ed874193993178cbf8f32c83aa2cbf9&vid=1cb223000ed874193993178cbf8f32c83aa2cbf9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR