Stearoyl coenzyme A

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Stearoyl Coenzyme A, Free Acid

19180010MG
100284-580EA 555.77 USD
100284-580
Stearoyl coenzyme A
Stearoyl coenzyme A
Formula: C₃₉H₇₀N₇O₁₇P₃S
MW: 1034.002 g/mol
MDL Number: MFCD00079568
CAS Number: 362-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR