Butyryl-L-carnitine chloride

Supplier: INDOFINE CHEMICAL COMPANY MS

17040025MG
100284-502EA 601.54 USD
100284-502
Butyryl-L-carnitine chloride
Butyryl-L-carnitine chloride
Formula: C₁₁H₂₂ClNO₄
MW: 267.75 g/mol
MDL Number: MFCD00274128
CAS Number: 162067-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f0677fd606e935cb2836d5228f64004da83921&vid=b0f0677fd606e935cb2836d5228f64004da83921&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR