1,11-Undecanedioic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1,9-Nonanedicarboxylic acid, 1,9-Nonanedicarboxylicacid, Nonamethylenedicarboxylic acid, Undecanedioic acid

151100100MG
100284-464EA 425 USD
100284-464
1,11-Undecanedioic acid
1,11-Undecanedioic acid
Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 372
MDL Number: MFCD00004444
CAS Number: 1852-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72aa6c92594e86c2516ee613061968169314208f&vid=72aa6c92594e86c2516ee613061968169314208f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR