2-Hydroxyphytanic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

1416425MG
100284-232EA 746.69 USD
100284-232
2-Hydroxyphytanic acid
2-Hydroxyphytanic acid
Formula: C₂₀H₄₀O₃
MW: 328.537 g/mol
MDL Number: MFCD05864053
CAS Number: 14721-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92c2ba27dff43423f2ab7fc22c63ff0390239eda&vid=92c2ba27dff43423f2ab7fc22c63ff0390239eda&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR