2-Hydroxy-n-octanoic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Hydroxy-n-octanoic Acid, 2-Hydroxyoctanoic acid

14080250MG
100284-174EA 545.31 USD
100284-174
2-Hydroxy-n-octanoic acid
2-Hydroxy-n-octanoic acid
Formula: C₈H₁₆O₃
MW: 160.21 g/mol
MDL Number: MFCD00014410
CAS Number: 617-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92812c3b713bfc4ac5a81526339ec7ea47c9a225&vid=92812c3b713bfc4ac5a81526339ec7ea47c9a225&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR