3-Hydroxyhexanoic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

14060325MG
100284-170EA 601.54 USD
100284-170
3-Hydroxyhexanoic acid
3-Hydroxyhexanoic acid
Formula: C₆H₁₂O₃
MW: 132.159 g/mol
MDL Number: MFCD02259045
CAS Number: 10191-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c42e64cb794300fb602b79bb3e9df5ca4256bb8&vid=9c42e64cb794300fb602b79bb3e9df5ca4256bb8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR