Methyl perfluorohexadecanoate ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

098895GM
100283-844EA 122.92 USD
100283-844
Methyl perfluorohexadecanoate ≥95%
Methyl perfluorohexadecanoate
Formula: C₁₇H₃F₃₁O₂
MDL Number: MFCD00236617
CAS Number: 165457-57-8
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd5b04f90b7f901f0a718837650f8664fe062539&vid=fd5b04f90b7f901f0a718837650f8664fe062539&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR