1-Fluorooctane ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Octyl fluoride

0935285GM
100283-404EA 204 USD
100283-404
1-Fluorooctane ≥98%
1-Fluorooctane
Formula: C₈H₁₇F
MW: 132.22
Boiling Pt: 130
Density: 0.814
MDL Number: MFCD00013582
CAS Number: 463-11-6
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b20525180b459685b07b6c3079cc90d049dd6046&vid=b20525180b459685b07b6c3079cc90d049dd6046&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR