1-Nitro-4-(trifluoromethoxy)benzene ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

09248225GM
100283-080EA 124.23 USD
100283-080
1-Nitro-4-(trifluoromethoxy)benzene ≥98%
1-Nitro-4-(trifluoromethoxy)benzene
Formula: C₇H₄F₃NO₃
MDL Number: MFCD00040302
CAS Number: 713-65-5
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef5a8506e7b8d713f1d741ac94687c28dcc6d9b7&vid=ef5a8506e7b8d713f1d741ac94687c28dcc6d9b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR